ContactUsLinkComponent
Immunoassay microplates, Protein L coated plates, Pierce™
Immunoassay microplates, Protein L coated plates, Pierce™
Catalog # PIER15190
undefined
Immunoassay microplates, Protein L coated plates, Pierce™
Catalog # PIER15190
Specifications
  • Packed:
    5 plates per pack
  • Colour:
    Clear
  • Description:
    96-well plates, Protein L coated, Polystyrene
  • Pk:
    5
Specifications
About this item
Pierce™ Protein L coated plates provide an alternative to direct, passive adsorption methods for immobilising antibodies for ELISA and other plate-based assay techniques.
  • Retain antibody activity, which can be lost when antibodies are immobilised by passive adsorption
  • Orient antibodies for maximum antigen-binding capacity
  • Immobilise antibodies for plate assays without prior purification
  • Ensure minimal variation (<5% well-to-well) from consistent coating
  • Plates are pre-blocked with SuperBlock™ blocking buffer to reduce non specific binding
  • Bind 2 to 5 pmol of antibody per well (based on tests with rabbit IgG)
  • Binding Property: Standard Capacity, Affinity; Detection Method: Colorimetric
Protein L binds to immunoglobulin kappa light chains without interfering with the antigen-binding site. Protein L binds to all classes of Ig (ie. IgG, IgM, IgA, IgE and IgD). Protein L also binds single chain variable fragments (scFv) and Fab fragments.