Hydrazide resin, UltraLink™
Catalog # PIER53149
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:10 ml
- Description:UltraLink™ Hydrazide resin
- Type:Liquid
Specifications
About this item
UltraLink™ Hydrazide resin is used to prepare affinity columns with antibodies or other glycolsylated proteins.
- Able to immobilise 1 to 10 mg of oxidized antibody or other glycoprotein per ml of resin
- Immobilise in as few as 30 minutes; achieve at least 90% coupling of most glycoproteins in less than 4 hours (efficiency depends on the amount and type of glycosylation)
- Conjugates only to purified glycoproteins whose sugar groups have been gently oxidised with periodate
- No stabilisation step is required