About this item
HyClone™ SFM4CHO media is a serum-free and protein-free formulation that contains no components of bovine origin and has been designed for high performance in a variety of culture vessels, including bioreactors.
- Metabolically designed for high cell yield and recombinant protein production
- Supports multiple CHO clones
- Allows for direct or sequential adaptation
- Supports growth in multiple cell culture systems
- Component traceability
- Manufactured under cGMP
HyClone™ SFM4CHO media has been formulated using a proprietary lipid complexing process for enhanced stability and growth promotion of various CHO cell lines. It has been designed to support the DHFR selection/amplification system and is also available without L-glutamine to support the GS gene expression system.