HA-Tag Magnetic IP/Co-IP Kit, Pierce™
Catalog # PIER88838
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1 KIT
- Description:HA-Tag Magnetic IP/Co-IP Kit, sufficient for 40 IP reactions using 25 µl of anti-HA magnetic beads
Specifications
About this item
The Pierce™ HA-Tag Magnetic IP/Co-IP Kit contains specific immunoaffinity magnetic beads and reagents to perform immunoprecipitation assays of HA fusion proteins or co-IP experiments using HA-tagged bait proteins.
- Covalently immobilised high-quality anti-HA monoclonal antibody enables high yields of immunoprecipitation products
- Stable, pre-blocked beads and specific antibody minimise off-target binding
- Complete kit and easy to follow instructions provide optimised protocols to perform IP or Co-IP experiments in approximately 1 hour
- Magnetic beads are compatible with manual and automated magnetic separation workflows
Delivery: The kit includes HA-tagged positive control , 500 µl; anti-HA magnetic beads, 1 ml; IP lysis/wash buffer, 2×50 ml; non-reducing sample buffer (5X), 5 ml; elution buffer (pH 2,0), 5 ml; neutralization buffer (pH 8,5), 1 ml.