Narcosis inhalation chamber, SP Bel-Art
Catalog # BELAH464200002
Supplier: Bel-Art Products, a Part of SP
Specifications
- Description:Clear acrylic inhalation narcosis chamber, Ø: 197 mm
- Pk:1
Specifications
About this item
Inhalation narcosis chamber with hinged cover.
- Cover and additional PP components not affected by anesthetic compounds
- Folding perforated platform removed to fill the bottom with cotton
- Small funnel to pour the anesthetic where it is absorbed
Designed for the observation of small animals during anesthetising.