Desalting columns, Zeba™, Pierce™
Supplier: Thermo Fisher Scientific
Certificates
About this item
These spin desalting columns are made of low protein binding PP and are compatible with a wide range of standard laboratory instruments and consumables. They provide >95% retention of salts and other small molecules.
- Higher protein recovery
- Minimal sample dilution
- No screening fractions for protein or waiting for protein to emerge by gravity-flow
- Easy-to-use with no cumbersome column preparation or equilibration
The Zeba 7K MWCO is recommended for processing proteins >7 kDa, and the Zeba 40K MWCO for processing proteins >40 kDa.
Caution: For research use only. Not for use in diagnostic procedures.