Cryopreservation media, HyClone™ HyCryo™
Catalog # SR30001.02
Supplier: HyClone products (Cytiva)
CAS Number:
Specifications
- Pk:100 ml
- Description:HyClone™ HyCryo cryopreservation medium
Specifications
About this item
Animal origin-free HyClone™ HyCryo cryopreservation media is designed for general use.
- 0,2 μm sterile filtered
- Serum free formulation
- Increased recovery of viable cells versus standard cryopreservation formulations