ContactUsLinkComponent
Manual microplate heat sealer, ALPS™ 30
Catalog # MATRAB-0400
   Low Manufacturing Impact
Manual microplate heat sealer, ALPS™ 30
Catalog # MATRAB-0400
   Low Manufacturing Impact
Specifications
  • Description:
    ALPS™ 30
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    NAR Certificate of Retirement
  • Pk:
    1
Specifications
About this item
Use of the ALPS™ 30 manual heat sealer protects samples from evaporation and contamination when performing PCR or sample storage. Perfect for sealing small numbers of plates in low to medium throughput applications, the ALPS™ 30 unit is small enough to keep on the bench and not take up a lot of space. Its simple, ergonomic manual operation provides quick, efficient sealing.
  • Compatible with virtually any height microplate or block, including PCR plates
  • Consistent heat seal provides dependable protection for PCR or sample storage plates
  • Prevents contamination and evaporation, yielding more reliable results
  • Features a built-in thermostat to prevent overheating
  • LED indicator lights show when power is on and when unit is heating
  • Small footprint so doesn't take up a lot of bench space, and lightweight so easy to carry
  • Three adapters included to accommodate most plate types: PCR plates, deep well plates and blocks, standard height 96- or 384-format plates