Ni-NTA magnetic beads, HisPur™, Pierce™
The Pierce™ Ni-NTA magnetic beads are high-capacity nickel-IMAC beads for affinity purification of His-tagged fusion proteins in manual or automated formats.
- Bead surface is pre-blocked and the protocol provides optimised buffers for purification
- Fast, scalable and versitle to purify proteins using native or denaturing conditions
- Can be use with common cell lysis reagents and a variety of buffer additives
- Protein coupling to the beads and downstream applications can be performed manually or by automation