Pierce™ Immunoassay Microplates, Antibody Coated, 96 Well
Supplier: Thermo Fisher Scientific
Certificates
About this item
Pierce™ Antibody coated plates are ideal for binding assays when available antibodies are in low quantities or they denature and become inactive upon direct adsorption to polystyrene plates. Clear plates for colorimetric ELISA and other assays. White plates for chemiluminescent. Black plates for fluorescent assays.
- Antibody-binding capacity is higher than direct adsorption onto polystyrene
- Ideal for direct, indirect, competitive or sandwich assays
- Activation level: 100 µl/well
- Pre-blocked with SuperBlock™ buffer to reduce non specific binding