ContactUsLinkComponent
Sigma-Aldrich® Ficoll® PM400
Sigma-Aldrich® Ficoll® PM400
Catalog # SIALF4375-50G
Supplier:  Merck
Sigma-Aldrich® Ficoll® PM400
Catalog # SIALF4375-50G
Supplier:  Merck
Specifications
  • Description:
    Ficoll® 400
  • Seq:
    03
  • Pk:
    50 g
Specifications
About this item
Ficoll® PM400 is a non-ionic synthetic polymer of sucrose used for cell separation and organelle isolation.
  • Concentrations of up to 50% (w/v) covering densities of up to 1,2 g/ml can be achieved
  • Better osmotic properties than sucrose
  • Preserves functional and morphological integrity
  • Does not penetrate biological membranes
  • Serves as the raw material for preparation of Ficoll-Paque gradients
Ficoll® is a registered trademark of GE Healthcare group of companies.