Ficoll® 400, BioXtra, lyophilised powder, for molecular biology, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
A non-ionic synthetic polymer of sucrose.
- Non-ionic polymer free of DNases, RNases, phosphatases and proteases
- Used for cell separation and organ isolation
- Separates cells with similar density but different size (PBMC from human peripheral blood, bone marrow and umbilical cord blood)
- Density ranges attained up to 1,2 g/ml
Ficoll® 400 polymer is known as synthetic polymer of sucrose which produce density gradients to separate subcellular components.