Order Entry
Germany
ContactUsLinkComponent
Ion selective electrode, half-cell, chloride, sensION+ 9652
Ion selective electrode, half-cell, chloride, sensION+ 9652
Catalog # HACHLZW9652970002
Supplier:  Hach
undefined
Ion selective electrode, half-cell, chloride, sensION+ 9652
Catalog # HACHLZW9652970002
Supplier:  Hach

Specifications

  • Concentration:
    1 mg/l (3×10-6 M) - 35500 mg/l (1 M) Cl-
  • Connection:
    BNC, fixed 1 m cable
  • Ø×L:
    12×120 mm
  • Type:
    sensION+ 9652
  • Description:
    Ion selective electrode (half-cell), chloride, sensION+ 9652, solid-state crystal membrane
  • Material:
    Epoxy
  • Pk:
    1
  • Temp.:
    5...50 °C

Specifications

About this item

sensION+ 9652 is a half-cell (reference not integrated) chloride ion selective electrode with an epoxy body and non-replaceable crystalline membrane selective to chloride ions in solution.

  • Utilises a unique solid-state sensor technology that eliminates costly membrane replacements
  • Requires virtually no maintenance: Solid-state sensor design allows for dry storage of the ISE without a shelf life or membrane replacement
  • Provides fast, stable, and accurate response in a variety of sample types

The 9652 has a fixed 1 meter cable and BNC connector for laboratory use with the Hach sensION+ MM374 Multimeter. The 9652 is ideal for measuring chloride concentrations in wastewater, drinking water and general water quality applications.

Delivery: Supplied with basic user manual.

Reference electrode

Product Information