Order Entry
Germany
ContactUsLinkComponent
EDTA disodium salt 2% in aqueous solution, Sigma-Aldrich®
EDTA disodium salt 2% in aqueous solution, Sigma-Aldrich®
Catalog # SIAL2854-15ML
Supplier:  Merck
CAS Number:  
EDTA disodium salt 2% in aqueous solution, Sigma-Aldrich®
Catalog # SIAL2854-15ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    15 mL
  • Seq:
    2295

Specifications

About this item

 :
 :336.21 g/mol
 :M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:205-358-3
CAS Number:139-33-3
Formula:C₁₀H₁₄N₂Na₂O₈
MDL Number:MFCD00070672