Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Staurosporine
Staurosporine ≥98%
Staurosporine ≥98%
Staurosporine ≥98%
Supplier:
MP Biomedicals
Certificates
Expand image
Staurosporine ≥98%
Supplier:
MP Biomedicals
About this item
CAS Number:
62996-74-1
:
Formula:
C₂₈H₂₆N₄O₃
MDL Number:
MFCD18252446
:
466.541 g/mol