Desmosine 99%, yellow solid
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:11003-57-9
:
Formula:C₂₄H₃₉N₅O₈
:526 g/mol (anhy)
Storage Temperature:Refrigerator
Specification Test Results
- Identity Test:Passes
- Source:Bovine Neck Ligament
- Purity:99%