Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
fenoterol
fenoterol, powder
fenoterol, powder
fenoterol, powder
Supplier:
MP Biomedicals
Certificates
Expand image
fenoterol, powder
Supplier:
MP Biomedicals
About this item
CAS Number:
13392-18-2
:
Formula:
C₁₇H₂₁NO₄
:
303.358 g/mol