Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methyl myristoleate
Methyl myristoleate ≥99%, liquid
Methyl myristoleate ≥99%, liquid
:
MP Biomedicals
Methyl myristoleate ≥99%, liquid
:
MP Biomedicals
:
56219-06-8
:
:
C₁₅H₂₈O₂
:
MFCD00056299
:
240.387 g/mol