Ryania
Catalog # ICNA0215377001
Supplier: MP Biomedicals
CAS Number:
Specifications
- Pk:1 mg
- Seq:2070
Specifications
Specification Test Results
- Identity Test:Passes
- Source:Ryania speciosa
Specification Test Results
About this item
:
:493.553 g/mol
CAS Number:15662-33-6
Formula:C₂₅H₃₅NO₉