Order Entry
Germany
ContactUsLinkComponent
 
β-Dihydronicotinamide adenine dinucleotide disodium salt (NADH-Na₂, reduced form) ≥98%
 
β-Dihydronicotinamide adenine dinucleotide disodium salt (NADH-Na₂, reduced form) ≥98%

 

  • Form:
    White Powder
  • Storage:
    Store at −0 °C. Store Desiccated. Store Under Nitrogen. Protect from light
  • pH:
    7,5 - 9,5 (10% aq soln)