Carboxymethyl cellulose sodium salt FCC, low viscosity
Catalog # SPCMC1103-2.5KG
Supplier: Spectrum Chemical
CAS Number:
Specifications
- Pack type:Poly bottle
- Pk:2,5 kg
- Seq:4076
Specifications
Specification Test Results
- Assay:99.5% Min.
Specification Test Results
About this item
:
Merck Index:12,01877
Melting Pt:300 °C
Storage Temperature:Ambient
CAS Number:9004-32-4
Formula:[C₆H₇O₂(OH)x(OCH₂COONa)y]n
MDL Number:MFCD00081472