0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Germany
ContactUsLinkComponent
Calcium acetate
Calcium acetate USP
Calcium acetate USP
:
Spectrum Chemical
Calcium acetate USP
:
Spectrum Chemical
:
62-54-4
:
:
(H₃CCOO)₂Ca
:
MFCD00150019
:
158.17 g/mol
:
12,01683
:
1.509 g/cm³ (20 °C)
:
Ambient
:
200-540-9
Assay:
99% Min.