Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Saccharin sodium dihydrate
Saccharin sodium dihydrate, granular USP
Saccharin sodium dihydrate, granular USP
:
Spectrum Chemical
Saccharin sodium dihydrate, granular USP
:
Spectrum Chemical
:
6155-57-3
:
:
C₇H₅NNaO₃S·2H₂O
:
MFCD00151213
:
242.21 g/mol
:
204-886-1
Assay:
99% Min.