Betaine hydrochloride USP
Supplier: Spectrum Chemical
Certificates
About this item
CAS Number:590-46-5
:
Formula:C₅H₁₂ClNO₂
MDL Number:MFCD00011903
:153.61 g/mol
Storage Temperature:Ambient
EINECS:209-683-1
Specification Test Results
- Assay:98% Min.