0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Germany
ContactUsLinkComponent
Aluminium chloride hexahydrate
Aluminium chloride hexahydrate USP
Aluminium chloride hexahydrate USP
:
Spectrum Chemical
Aluminium chloride hexahydrate USP
:
Spectrum Chemical
:
7784-13-6
:
:
AlCl₃·6H₂O
:
MFCD00149134
:
241.43 g/mol
:
∼100 °C
:
2.39 g/cm³ (20 °C)
:
Ambient
:
231-208-1
:
3260
:
8,III
:
GHS07
Assay:
95% Min.