Order Entry
Germany
Orders LinkContactUsLinkComponent
Lanolin USP

Lanolin USP

Lanolin USP
Supplier:  Spectrum Chemical
Certificates

2 Options Available Below

Product Details & Documents
Synonyms:  Wool fat
  • CAS Number:8006-54-0
  • MDL Number:MFCD00081740
  • Merck Index:12,05371
  • Melting Pt:38…44 °C
  • Density:0.932…0.945 g/cm³ (15 °C)
  • Flash Pt:113 °C (closed cup)
  • EINECS:232-348-6

Documents

Product Family Options

Product Information

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.