Diethylenetriaminepentaacetic acid
Supplier: Spectrum Chemical
Certificates
About this item
CAS Number:67-43-6
:
Formula:C₁₄H₂₃N₃O₁₀
MDL Number:MFCD00004289
Merck Index:13,07202
Melting Pt:220…222 °C
Density:1.558 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-652-8
UN:3077
ADR:9,III
:393.35 g/mol
:GHS07
Specification Test Results
- Assay:98% Min.