Order Entry
Germany
Orders LinkContactUsLinkComponent
L(+)-Cysteine

L(+)-Cysteine

Irritant
Supplier:   Spectrum Chemical
L(+)-Cysteine
Supplier:  Spectrum Chemical
Certificates

3 Options Available Below

Product Details & Documents
Synonyms:  H-Cys-OH, L-(+)-Cysteine, (R)-2-Amino-3-mercaptopropanoic acid
  • CAS Number:52-90-4
  • Formula:HSCH₂CH(NH₂)CO₂H
  • MDL Number:MFCD00064306
  • Molecular Weight:121.16 g/mol
  • Merck Index:13,02810
  • Boiling Pt:254 °C (1013 hPa)
  • Melting Pt:115 °C
  • Density:1.523 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-158-2
Specifications for Product Family

Specification Test Results

  • Assay:
    98% Min.
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass, amber
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg - 754,00 €
    754,00 €
    1 * 1 kg
       754,00 €

  • Fiber drum
    25 kg
    Log in for availability
    Log in for availability
    1 * 25 kg
    1 * 25 kg

  • 50 kg
    Log in for availability
    Log in for availability
    1 * 50 kg
    1 * 50 kg - 13 150,00 €
    13 150,00 €
    1 * 50 kg
       13 150,00 €