(3S)-(-)-3-(tert-Butoxycarbonylamino)pyrrolidine 99%, ee 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:122536-76-9
:
Formula:C₉H₁₈N₂O₂
MDL Number:MFCD00143194
:186.26 g/mol
Melting Pt:50…54 °C
Storage Temperature:Refrigerator
UN:3259
ADR:8,III
:GHS05