Order Entry
Germany
ContactUsLinkComponent
tert-Butyl pyrrolidine-1-carboxylate 98%
tert-Butyl pyrrolidine-1-carboxylate 98%
Catalog # L17475.06
CAS Number:  
tert-Butyl pyrrolidine-1-carboxylate 98%
Catalog # L17475.06
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

CAS Number:86953-79-9
 :
Formula:C₉H₁₇NO₂
MDL Number:MFCD00216581
 :171.24 g/mol
Boiling Pt:130 °C (20 mmHg)
Density:0.977
Flash Pt:86 °C (186 °F)