
Magnesium bromide diethyl etherate 98%
Catalog # L15481.14
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
About this item
CAS Number:29858-07-9
:
Formula:C₄H₁₀Br₂MgO
:258.236 g/mol
Flash Pt:35 °C (95 °F)
Storage Temperature:Ambient
ADR:4.1,II
:GHS02
MDL Number:MFCD00064500