Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Sodium-tert-pentyloxide
Sodium-tert-pentyloxide 95%
Sodium-tert-pentyloxide 95%
Sodium-tert-pentyloxide 95%
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
Sodium-tert-pentyloxide 95%
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
14593-46-5
:
Formula:
C₅H₁₁NaO
MDL Number:
MFCD00064245
:
110.132 g/mol
EINECS:
238-369-4
ADR:
4.2,II
:
GHS02
:
GHS05
Storage Temperature:
Ambient
Related Information
Documentation
Certificates
Search by Lot Number
SDS
msds_ALFAL15112
Videos
Thermo Scientific Acroseal 25 Years
Documentation