
2,3-Dihydro-1H-inden-2-amine hydrochloride 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2338-18-3
:
Formula:C₉H₁₂ClN
MDL Number:MFCD00012549
:169.65 g/mol
Melting Pt:243…248 °C
EINECS:219-048-0
:GHS07
Storage Temperature:Ambient