
(S)-(+)-2-Dibenzylamino-3-phenyl-1-propanol 99%
Catalog # L13916.06
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
About this item
CAS Number:111060-52-7
:
Formula:C₂₃H₂₅NO
MDL Number:MFCD00191984
:331.46 g/mol
Melting Pt:72…74 °C
:GHS07
Storage Temperature:Ambient