
N-Acetylethylenediamine 90%, tech.
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1001-53-2
:
Formula:C₄H₁₀N₂O
MDL Number:MFCD00008163
:102.14 g/mol
Boiling Pt:128 °C (3 mmHg)
Melting Pt:44…50 °C
Flash Pt:>110 °C (230 °F)
Storage Temperature:Ambient
UN:3259
ADR:8,III
:GHS05