
Dodecylamine hydrochloride 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:929-73-7
:
Formula:C₁₂H₂₇N
MDL Number:MFCD00042017
Melting Pt:186…188 °C
:221.81 g/mol
EINECS:213-205-7
:GHS07
Storage Temperature:Ambient