4-Aminostyrene 97% stabilised
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1520-21-4
:
Formula:C₈H₉N
MDL Number:MFCD00015329
:119.17 g/mol
Boiling Pt:92…94 °C (4 mmHg)
Melting Pt:23…24 °C
Density:1.014
Flash Pt:>110 °C (230 °F)
EINECS:216-185-8
:GHS08
:GHS07