3-Benzylidene-2,4-pentanedione 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:4335-90-4
:
Formula:C₁₂H₁₂O₂
MDL Number:MFCD00014990
:188.23 g/mol
Boiling Pt:185…188 °C (15 mmHg)
Density:1.093
Flash Pt:>110 °C (230 °F)
Storage Temperature:Ambient
EINECS:224-382-5