Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Platinum
Platinum 10% on carbon, dry
Platinum 10% on carbon, dry
:
Thermo Fisher Scientific
Platinum 10% on carbon, dry
:
Thermo Fisher Scientific
:
7440-06-4
:
:
Pt
:
MFCD00011179
:
195.08 g/mol
:
14,7529
:
Ambient
:
231-116-1
:
1325
:
4.1,III
:
GHS02
msds_ALFAL00469
Thermo Scientific Acroseal 25 Years