Bisacrylamide 99+% for electrophoresis
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:110-26-9
:
Formula:(H₂C=CHCONH)₂CH₂
MDL Number:MFCD00008625
:154.17 g/mol
Melting Pt:>300 °C
Density:1.235
EINECS:203-750-9
:GHS07
Storage Temperature:Ambient