1-(((6R,7R)-7-((Z)-2-(2-Aminothiazol-4-yl)-2-(methoxyimino)acetamido)-2-carboxy-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-en-3-yl)methyl)-6,7-dihydro-5H-cyclopenta[b]pyridin-1-ium hydrogensulphate 95%
Catalog # J66001.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1 g
Specifications
About this item
:
:612.66 g/mol
Melting Pt:198…202 °C
CAS Number:98753-19-6
Formula:C₂₂H₂₄N₆O₉S₃
MDL Number:MFCD11864966