Carbonate-buffered saline (5X), pH 9,5
Catalog # J63928.AP
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:500 mL
- CAS No.:497-19-8
- Description:Carbonate-buffered Salin, 5X
- MDL No.:MFCD00003494
- Physical form:Liquid
- pH value:9.5
Specifications
About this item
CAS No.: 497-19-8
Physical Form: Liquid
MDL No.: MFCD00003494