Order Entry
Germany
ContactUsLinkComponent
Triton® X-100 lysis buffer with glycerol (2X), pH 7,4
Triton® X-100 lysis buffer with glycerol (2X), pH 7,4
Catalog # J63866.AK
CAS Number:  
Triton® X-100 lysis buffer with glycerol (2X), pH 7,4
Catalog # J63866.AK
CAS Number:  

Specifications

  • Pk:
    250 mL
  • Description:
    Triton® X-100 lysis buffer with glycerol (2X), pH 7.4
  • Physical form:
    Liquid
  • Appearance:
    Colourless

Specifications

About this item

Physical Form: Liquid
Appearance: Colourless