Citrate-buffered saline (20X)
Supplier: Thermo Fisher Scientific
Certificates
About this item
Saline-sodium citrate (SSC) buffer.
300 mM sodium citrate, 3M sodium chloride, pH 7.0
Specifications
- MDL No.:MFCD00132371
- Physical form:Liquid
Saline-sodium citrate (SSC) buffer.
300 mM sodium citrate, 3M sodium chloride, pH 7.0