BCA protein assay kit
Catalog # J63283.QA
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1 KIT
- Description:BCA protein assay kit
Specifications
About this item
BCA Protein Assay Kit can be used to measure very dilute protein concentrations in very small sample volumes. The kit accurately measures protein concentrations from 0,5 to 30 μg/ml in tube assays and 1 to 20 μg/ml in 96- or 384-well plate assays. It is used for studying protein:protein interactions, measuring column fractions after affinity chromatography, estimating percent recovery of membrane proteins from cell extracts and high-throughput screening of fusion proteins.
- Clear liquid
Physical Form: Liquid Appearance: Clear liquid
Applications: Contains two separate solutions for estimation of protein concentration.
Delivery: Contains 250 ml of Solution A (Bicinchonic acid and tartrate in an alkaline carbonate buffer) and 5,5 ml of Solution B (4% w/v cupric sulfate).