Fendiline hydrochloride
Catalog # J63254.06
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:351.92 g/mol
Merck Index:14,3970
Melting Pt:204…205 °C
EINECS:237-121-5
:GHS07
Storage Temperature:Ambient
CAS Number:13636-18-5
Formula:C₂₃H₂₅N·HCl
MDL Number:MFCD00079301