Order Entry
Germany
ContactUsLinkComponent
DNA marker, high range, 1000 bp ladder
DNA marker, high range, 1000 bp ladder
Certificates
DNA marker, high range, 1000 bp ladder
Certificates

Specifications

  • Form:
    Liquid
  • Storage temperature:
    -30°C to -10°C

Specifications

Related Information