Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Flunarizine dihydrochloride
Flunarizine dihydrochloride 99%
Flunarizine dihydrochloride 99%
J62969.03
:
Thermo Fisher Scientific
:
Flunarizine dihydrochloride 99%
J62969.03
:
Thermo Fisher Scientific
:
Pk:
1 g
Seq:
3047
:
:
477.40 g/mol
:
14,4144
:
250-216-6
:
GHS07
:
30484-77-6
:
C₂₆H₂₆F₂N₂·2HCl
:
MFCD00058198