Order Entry
Germany
ContactUsLinkComponent
 
MES lysis buffer with Triton® X-100
  J62693.AK
 :  
MES lysis buffer with Triton® X-100
  J62693.AK
 :  

 

  • Pk:
    250 mL
  • Description:
    MES lysis buffer with Triton® X-100
  • Storage temperature:
    2°C to 8°C
  • Physical form:
    Liquid
  • Appearance:
    Colourless

 

 

Storage Temperature: 2 to 8 °C
Physical Form: Liquid
Appearance: Colourless