Order Entry
Germany
ContactUsLinkComponent
TRIS-buffered saline (TBS, 10X) pH 7,6
TRIS-buffered saline (TBS, 10X) pH 7,6
Certificates
TRIS-buffered saline (TBS, 10X) pH 7,6
Certificates

Specifications

  • CAS No.:
    77-86-1
  • MDL No.:
    MFCD00004679
  • Physical form:
    Liquid

Specifications

Related Information